Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® 514)
HP-3G10 is an anti-human monoclonal antibody that recognizes the CD161 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
: For research use only.
Type: Secondary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® 514
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- 77024-596
- 77024-864
- Antigen symbol:
- CD161
- CD161
- Antigen name:
- CD161 antigen
- CD161 antigen
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 514
- iFluor® 514
- Clone:
- HP-3G10
- HP-3G10
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Purification:
- Affinity purification
- Affinity purification
- Storage temperature:
- Refrigerated (2...8 °C); Minimize light exposure
- Refrigerated (2...8 °C); Minimize light exposure
- Shipping temperature:
- 20
- 20
Frequently Bought Together