Order Entry
Canada
ContactUsLinkComponent
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PacOrange)
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PacOrange)
Supplier:  AAT BIOQUEST INC
Certificates
undefined
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (PacOrange)
Supplier:  AAT BIOQUEST INC

Specifications

  • Catalog No:
  • 77021-856
  • 77022-076
  • Antigen symbol:
  • CD44
  • CD44
  • Antigen name:
  • CD44 antigen
  • CD44 antigen
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Pacific Orange
  • Pacific Orange
  • Clone:
  • IM7
  • IM7
  • Reactivity:
  • Human
  • Human
  • Host:
  • Rat
  • Rat
  • Purification:
  • Affinity purification
  • Affinity purification
  • Storage temperature:
  • Refrigerated (2...8 °C); Minimize light exposure
  • Refrigerated (2...8 °C); Minimize light exposure
  • Shipping temperature:
  • 20
  • 20

Specifications

Frequently Bought Together