Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (APC/iFluor® 750)
Supplier: AAT BIOQUEST INC
Certificates
About this item
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Caution: For research use only.
Type: Secondary
Antigen: CD44
Clonality: Monoclonal
Clone: IM7
Conjugation: APC/iFluor® 750
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- 77023-938
- 77024-432
- 77021-612
- Antigen symbol:
- CD44
- CD44
- CD44
- Antigen name:
- CD44 antigen
- CD44 antigen
- CD44 antigen
- Antibody type:
- Secondary
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)/iFluor® 750
- APC (Allophycocyanin)/iFluor® 750
- APC (Allophycocyanin)/iFluor® 750
- Clone:
- IM7
- IM7
- IM7
- Reactivity:
- Human
- Human
- Human
- Host:
- Rat
- Rat
- Rat
- Purification:
- Affinity purification
- Affinity purification
- Affinity purification
- Storage temperature:
- Refrigerated (2...8 °C); Minimize light exposure
- Refrigerated (2...8 °C); Minimize light exposure
- Refrigerated (2...8 °C); Minimize light exposure
- Shipping temperature:
- 20
- 20
- 20
Specifications
Frequently Bought Together