Order Entry
Canada
ContactUsLinkComponent
 
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (mFluor™ Violet)
 
undefined
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (mFluor™ Violet)

 

  • Catalog No:
  • 77024-288
  • 77023-424
  • Antigen symbol:
  • CD38
  • CD38
  • Antigen name:
  • CD38 antigen
  • CD38 antigen
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • mFluor™ Violet
  • mFluor™ Violet
  • Clone:
  • HIT2
  • HIT2
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Purification:
  • Affinity purification
  • Affinity purification
  • Storage temperature:
  • Refrigerated (2...8 °C); Minimize light exposure
  • Refrigerated (2...8 °C); Minimize light exposure
  • Shipping temperature:
  • 20
  • 20

 

Frequently Bought Together