Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (iFluor® 546)
Supplier: AAT BIOQUEST INC
Certificates
About this item
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Caution: For research use only.
Type: Secondary
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: iFluor® 546
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Specifications
- Catalog No:
- 77023-276
- 77022-760
- Antigen symbol:
- CD38
- CD38
- Antigen name:
- CD38 antigen
- CD38 antigen
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 546
- iFluor® 546
- Clone:
- HIT2
- HIT2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Purification:
- Affinity purification
- Affinity purification
- Storage temperature:
- Refrigerated (2...8 °C); Minimize light exposure
- Refrigerated (2...8 °C); Minimize light exposure
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together