Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® A7)
4H11 is an anti-human monoclonal antibody that recognizes the CD34 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
: For research use only.
Type: Secondary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: iFluor® A7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
- Catalog No:
- 77024-008
- 77021-374
- Antigen symbol:
- CD34
- CD34
- Antigen name:
- CD34 antigen
- CD34 antigen
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® A7
- iFluor® A7
- Clone:
- 4H11
- 4H11
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1
- Purification:
- Affinity purification
- Affinity purification
- Storage temperature:
- Refrigerated (2...8 °C); Minimize light exposure
- Refrigerated (2...8 °C); Minimize light exposure
- Shipping temperature:
- 20
- 20
Frequently Bought Together