Order Entry
Canada
ContactUsLinkComponent
 
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 633)
 
undefined
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 633)

 

  • Catalog No:
  • 77024-502
  • 77022-392
  • Antigen symbol:
  • CD34
  • CD34
  • Antigen name:
  • CD34 antigen
  • CD34 antigen
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • iFluor® 633
  • iFluor® 633
  • Clone:
  • 4H11
  • 4H11
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • Purification:
  • Affinity purification
  • Affinity purification
  • Storage temperature:
  • Refrigerated (2...8 °C); Minimize light exposure
  • Refrigerated (2...8 °C); Minimize light exposure
  • Shipping temperature:
  • 20
  • 20

 

Frequently Bought Together