Order Entry
Canada
ContactUsLinkComponent
Sodium dodecyl sulfate (SDS) 20% in aqueous solution, OmniPur®, Millipore®
Sodium dodecyl sulfate (SDS) 20% in aqueous solution, OmniPur®, Millipore®
Catalog # CA-EM7990
Supplier:  MilliporeSigma
CAS Number:  
undefined
Sodium dodecyl sulfate (SDS) 20% in aqueous solution, OmniPur®, Millipore®
Catalog # CA-EM7990
Supplier:  MilliporeSigma
Supplier Number:  7990-200ML
CAS Number:  

Specifications

  • Pack type:
    Poly bottle
  • Size:
    200 ml
  • Cat. no.:
    CA-EM7990
  • Supplier No.:
    7990

Specifications

Specification Test Results

  • Max. UV Absorbance at 260 nm:
    0.1
  • Max. UV Absorbance at 280 nm:
    0.1
  • Conductivity:
    21,900–25,500 µmhos
  • pH (at 25°C):
    5.0–8.0
  • Phosphate (PO4):
    0.0005% max.

Specification Test Results

About this item

 :
 :288.38 g/mol
Density:1…1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-788-1
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175