Anti-CD28 Mouse Monoclonal Antibody (iFluor® 532) [clone: CD28.2]
Supplier: AAT BIOQUEST INC
Certificates
About this item
CD28.2 is an anti-human monoclonal antibody that forms an immune complex with the CD28 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Caution: For research use only.
Type: Secondary
Antigen: CD28
Clonality: Monoclonal
Clone: CD28.2
Conjugation: iFluor® 532
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- 77021-388
- 77024-088
- Antigen symbol:
- CD28
- CD28
- Antigen name:
- CD28 antigen
- CD28 antigen
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 532
- iFluor® 532
- Clone:
- CD28.2
- CD28.2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Purification:
- Affinity purification
- Affinity purification
- Storage temperature:
- Refrigerated (2...8 °C); minimize light exposure
- Refrigerated (2...8 °C); minimize light exposure
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together