Order Entry
Canada
ContactUsLinkComponent
Anti-CD19 Mouse Monoclonal Antibody (mFluor™ Red) [clone: SJ25C1]
Anti-CD19 Mouse Monoclonal Antibody (mFluor™ Red) [clone: SJ25C1]
Supplier:  AAT BIOQUEST INC
Certificates
undefined
Anti-CD19 Mouse Monoclonal Antibody (mFluor™ Red) [clone: SJ25C1]
Supplier:  AAT BIOQUEST INC

Specifications

  • Catalog No:
  • 77021-134
  • 77022-022
  • Antigen symbol:
  • CD19
  • CD19
  • Antigen name:
  • CD19 antigen
  • CD19 antigen
  • Antibody type:
  • Secondary
  • Secondary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • mFluor™ Red
  • mFluor™ Red
  • Clone:
  • SJ25C1
  • SJ25C1
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Purification:
  • Affinity purification
  • Affinity purification
  • Storage temperature:
  • Refrigerated (2...8 °C); minimize light exposure
  • Refrigerated (2...8 °C); minimize light exposure
  • Shipping temperature:
  • 20
  • 20

Specifications

Frequently Bought Together