Anti-CD19 Mouse Monoclonal Antibody (iFluor® 450) [clone: SJ25C1]
Supplier: AAT BIOQUEST INC
Certificates
About this item
SJ25C1 is an anti-human monoclonal antibody that forms an immune complex with the CD19 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Caution: For research use only.
Type: Secondary
Antigen: CD19
Clonality: Monoclonal
Clone: SJ25C1
Conjugation: iFluor® 450
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- 77023-300
- 77024-670
- Antigen symbol:
- CD19
- CD19
- Antigen name:
- CD19 antigen
- CD19 antigen
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 450
- iFluor® 450
- Clone:
- SJ25C1
- SJ25C1
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Purification:
- Affinity purification
- Affinity purification
- Storage temperature:
- Refrigerated (2...8 °C); minimize light exposure
- Refrigerated (2...8 °C); minimize light exposure
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together