Anti-CD16a Mouse Monoclonal Antibody (iFluor® 350) [clone: 3G8]
Supplier: AAT BIOQUEST INC
Certificates
About this item
The 3G8 monoclonal antibody recognizes human CD16a, a 50 to 65 kD transmembrane protein typically found on the surface of macrophages, natural killer cells, dendritic cells, monocytes and neutrophils.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Caution: For research use only.
Type: Secondary
Antigen: CD16a
Clonality: Monoclonal
Clone: 3G8
Conjugation: iFluor® 350
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- 77021-546
- 77024-546
- Antigen symbol:
- CD16a
- CD16a
- Antigen name:
- CD16a antigen
- CD16a antigen
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 350
- iFluor® 350
- Clone:
- 3G8
- 3G8
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Purification:
- Affinity purification
- Affinity purification
- Storage temperature:
- Refrigerated (2...8 °C); minimize light exposure
- Refrigerated (2...8 °C); minimize light exposure
- Shipping temperature:
- 20
- 20
Specifications
Frequently Bought Together