Anti-CD14 Mouse Monoclonal Antibody (iFluor® 670) [clone: 61D3]
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
: For research use only.
Type: Secondary
Antigen: CD14
Clonality: Monoclonal
Clone: 61D3
Conjugation: iFluor® 670
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
- Catalog No:
- 77024-034
- 77023-922
- Antigen symbol:
- CD14
- CD14
- Antigen name:
- CD14 antigen
- CD14 antigen
- Antibody type:
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- iFluor® 670
- iFluor® 670
- Clone:
- 61D3
- 61D3
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Purification:
- Affinity purification
- Affinity purification
- Storage temperature:
- Refrigerated (2...8 °C); minimize light exposure
- Refrigerated (2...8 °C); minimize light exposure
- Shipping temperature:
- 20
- 20
Frequently Bought Together