Order Entry
Canada
ContactUsLinkComponent
Anti-CD5 Mouse Monoclonal Antibody (PE/iFluor® 594) [clone: L17F12]
Fournisseur:  AAT BIOQUEST INC
Certificats
undefined
Anti-CD5 Mouse Monoclonal Antibody (PE/iFluor® 594) [clone: L17F12]
Fournisseur:  AAT BIOQUEST INC

Spécifications

  • Catalog No:
  • 77022-966
  • 77023-278
  • 77024-316
  • Antigen symbol:
  • CD5
  • CD5
  • CD5
  • Antigen name:
  • CD5 antigen
  • CD5 antigen
  • CD5 antigen
  • Antibody type:
  • Secondary
  • Secondary
  • Secondary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)/iFluor® 594
  • PE (Phycoerythrin)/iFluor® 594
  • PE (Phycoerythrin)/iFluor® 594
  • Clone:
  • L17F12
  • L17F12
  • L17F12
  • Reactivity:
  • Human
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Mouse
  • Purification:
  • Affinity purification
  • Affinity purification
  • Affinity purification
  • Storage temperature:
  • Refrigerated (2...8 °C); minimize light exposure
  • Refrigerated (2...8 °C); minimize light exposure
  • Refrigerated (2...8 °C); minimize light exposure
  • Shipping temperature:
  • 20
  • 20
  • 20

Spécifications

Frequently Bought Together