Anti-CD5 Mouse Monoclonal Antibody (APC/iFluor® 700) [clone: L17F12]
Supplier: AAT BIOQUEST INC
Certificates
About this item
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
- Purified through affinity chromatography
Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.
Caution: For research use only.
Type: Secondary
Antigen: CD5
Clonality: Monoclonal
Clone: L17F12
Conjugation: APC/iFluor® 700
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Specifications
- Catalog No:
- 77021-270
- 77021-892
- 77024-574
- Antigen symbol:
- CD5
- CD5
- CD5
- Antigen name:
- CD5 antigen
- CD5 antigen
- CD5 antigen
- Antibody type:
- Secondary
- Secondary
- Secondary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)/iFluor® 700
- APC (Allophycocyanin)/iFluor® 700
- APC (Allophycocyanin)/iFluor® 700
- Clone:
- L17F12
- L17F12
- L17F12
- Reactivity:
- Human
- Human
- Human
- Host:
- Mouse
- Mouse
- Mouse
- Purification:
- Affinity purification
- Affinity purification
- Affinity purification
- Storage temperature:
- Refrigerated (2...8 °C); minimize light exposure
- Refrigerated (2...8 °C); minimize light exposure
- Refrigerated (2...8 °C); minimize light exposure
- Shipping temperature:
- 20
- 20
- 20
Specifications
Frequently Bought Together