Orders LinkContactUsLinkComponent
Wallcur® Practi-Ipratropium Bromide Nebules
Wallcur® Practi-Ipratropium Bromide Nebules
Practi-ipratropium bromide 0.02% nebules. (0.5 mg/2.5 ml), Distilled water
Catalog # 470355-072
Supplier:   WALLCUR, LLC.
Supplier #:  1206PN
Wallcur® Practi-Ipratropium Bromide Nebules
Catalog # 470355-072
Supplier:  WALLCUR, LLC.
Wallcur® Practi-Ipratropium Bromide Nebules
Wallcur® Practi-Ipratropium Bromide Nebules
Practi-ipratropium bromide 0.02% nebules. (0.5 mg/2.5 ml), Distilled water
Price
$84.93
Log in for availability
Box of 30
Product Details & Documents
Made for training.
  • Contains no medication
  • Contents are safe for use with most simulators
Practi-Ipratropium Bromide™ 0.02% nebules simulates 0.5 mg/2.5 ml of ipratropium bromide 0.02% (Atrovent®) in a nebulizer. Practi-ipratropium comes as a liquid solution that is to be inhaled by the patient's mouth using a special nebulizer. Safely teach this skill using Wallcur's Practi-ipratropium bromide 0.02% with any task trainer or manikin that accepts distilled water. Students can also practice dosage calculations to understand the different dosages for adults versus pediatrics.
Delivery information: These products require a release form for purchase. If your account does not have one on file, the item cannot ship until release documentation is received.
Caution: For training purposes only. Not for human or animal use.
Specifications
  • Description:
    Practi-ipratropium bromide 0.02% nebules. (0.5 mg/2.5 ml)
  • Includes:
    Distilled water