Orders LinkContactUsLinkComponent
NEXTFLEX™ UDI Barcodes
NEXTFLEX™ UDI Barcodes
Certificates
NEXTFLEX™ UDI Barcodes
NEXTFLEX™ UDI Barcodes
6 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
The NEXTFLEX® Unique dual index barcodes are barcoded adapters that can be used with single or paired-end read sequencing library prep kits  designed for sequencing on Illumina® platforms workflows.
  • 384 UDI adapters for mitigating sample misassignment due to index hopping
  • Unique dual index adapters for multiplexing up to 384 libraries for Illumina® sequencing platforms
  • Mitigate index hopping or spread of signal that can occur during sequencing
  • Each lot of the NEXTFLEX® Unique dual index barcodes is tested for index purity by sequencing
  • Compatible with both paired-end and single-read sequencing
  • Compatible with the NEXTFLEX® Universal blockers
The NEXTFLEX® Unique dual index barcodes utilize two unique 8 nt indexes on each UDI adapter. The 8 nt indices are designed with a Hamming Distance of at least three throughout the entire series, permitting error correction. Patterned flow cells, such as those found on the Illumina® NovaSeq® and HiSeq® 3000/4000 platforms have been recognized to suffer from increased rates of sample mis-assignment during sequencing.
The use of adapters with unique dual indices during sequencing prevents such mis-assigned reads from appearing in final data sets, allowing for the highest assurance of data integrity. The NEXTFLEX® Unique dual index barcodes are barcoded adapters, for sequencing on Illumina® platforms, that provide data security in sequencing applications. Multiplexing with NEXTFLEX® Unique dual index barcodes increase scale while reducing costs by allowing the user to pool up to 384 libraries in a single flow cell lane.
 : Manufactured in ISO 9001 facility.
 : For research use only. Not for use in diagnostic procedures.
Product Family Options
Product Information
  • No. of reactions
    Availability
    Price
  • CA77448-164
    NEXTFLEX™ UDI barcodes 16 - Eval (1 - 8)
    NEXTFLEX™ UDI barcodes 16 - Eval (1 - 8)
    16
    Log in for availability
    Log in for availability
    Each
    Each - $249.04
    $249.04
    Each
       $249.04
    Supplier Number: NOVA-514150-EVAL16
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • CA77448-166
    NEXTFLEX™ UDI barcodes 48 - Eval (1 - 24)
    NEXTFLEX™ UDI barcodes 48 - Eval (1 - 24)
    48
    Log in for availability
    Log in for availability
    Each
    Each - $747.80
    $747.80
    Each
       $747.80
    Supplier Number: NOVA-514150-EVAL48
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • CA77448-162
    NEXTFLEX™ UDI barcodes (1 - 96)
    NEXTFLEX™ UDI barcodes (1 - 96)
    192
    Log in for availability
    Log in for availability
    Each
    Each - $3,035.98
    $3,035.98
    Each
       $3,035.98
    Supplier Number: NOVA-514150
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • CA77448-168
    NEXTFLEX™ UDI barcodes (97 - 192)
    NEXTFLEX™ UDI barcodes (97 - 192)
    192
    Log in for availability
    Log in for availability
    Each
    Each - $3,035.98
    $3,035.98
    Each
       $3,035.98
    Supplier Number: NOVA-514151
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • CA77448-170
    NEXTFLEX™ UDI barcodes (193 - 288)
    NEXTFLEX™ UDI barcodes (193 - 288)
    192
    Log in for availability
    Log in for availability
    Each
    Each - $3,035.98
    $3,035.98
    Each
       $3,035.98
    Supplier Number: NOVA-514152
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • CA77448-172
    NEXTFLEX™ UDI barcodes (289 - 384)
    NEXTFLEX™ UDI barcodes (289 - 384)
    192
    Log in for availability
    Log in for availability
    Each
    Each - $3,035.98
    $3,035.98
    Each
       $3,035.98
    Supplier Number: NOVA-514153
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.