Order Entry
Canada
Orders LinkContactUsLinkComponent
Fletcher's Medium Broth Base

Fletcher's Medium Broth Base

Rabbit serum, 125 ml, Bottle, screw cap
Catalog # 10052-852
Supplier:   Hardy Diagnostics
Supplier #:  SRS125
Fletcher's Medium Broth Base
Catalog # 10052-852
Supplier:  Hardy Diagnostics
CAS Number:  
Fletcher's Medium Broth Base
Fletcher's Medium Broth Base

Rabbit serum, 125 ml, Bottle, screw cap

CAS Number:  
Price
350,00 $
Log in for availability
Each (125ml)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

For the culture of Leptospira.

Ordering information: Supplement required, rabbit serum.

Specifications

Specifications

  • Pack type:
    Bottle, screw cap
  • Description:
    Rabbit serum
  • Volume:
    125 ml

Specifications