Orders LinkContactUsLinkComponent
Synthware Round-Bottom Flasks with Four Necks
Supplier:   Kemtech America
Synthware Round-Bottom Flasks with Four Necks
Supplier:  Kemtech America
Certificates
Synthware Round-Bottom Flasks with Four Necks
Synthware Round-Bottom Flasks with Four Necks
13 Options Available Below
Product Details & Documents
With identical vertical side joints.
  • 3 Straight side necks
  • Ground glass joints
  • Heavy wall borosilicate construction
  • Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
Product Family Options
Product Information
  • Capacity
    Center neck [ST] joint
    Side neck ST joints
    Availability
    Price
  • 250 ml
    24/40
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $277.03
    $277.03
    New Customer Promotional Price
    Each
       $277.03
    Supplier Number: F214250
  • 500 ml
    24/40
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $328.80
    $328.80
    New Customer Promotional Price
    Each
       $328.80
    Supplier Number: F214500
  • 500 ml
    29/42
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $289.41
    $289.41
    New Customer Promotional Price
    Each
       $289.41
    Supplier Number: F212500
  • 500 ml
    29/42
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $333.49
    $333.49
    New Customer Promotional Price
    Each
       $333.49
    Supplier Number: F218500
  • 1000 ml
    24/40
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $405.07
    $405.07
    New Customer Promotional Price
    Each
       $405.07
    Supplier Number: F214000
  • 1000 ml
    29/42
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $356.45
    $356.45
    New Customer Promotional Price
    Each
       $356.45
    Supplier Number: F21841L
  • 2000 ml
    29/42
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $405.20
    $405.20
    New Customer Promotional Price
    Each
       $405.20
    Supplier Number: F21842L
  • 2000 ml
    34/45
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $418.33
    $418.33
    New Customer Promotional Price
    Each
       $418.33
    Supplier Number: F21342L
  • 3000 ml
    45/50
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $583.27
    $583.27
    New Customer Promotional Price
    Each
       $583.27
    Supplier Number: F21543L
  • 3000 ml
    45/50
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $620.78
    $620.78
    New Customer Promotional Price
    Each
       $620.78
    Supplier Number: F21583L
  • 5000 ml
    29/42
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $804.65
    $804.65
    New Customer Promotional Price
    Each
       $804.65
    Supplier Number: F21845L
  • 5000 ml
    45/50
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $861.35
    $861.35
    New Customer Promotional Price
    Each
       $861.35
    Supplier Number: F21545L
  • 5000 ml
    45/50
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $869.75
    $869.75
    New Customer Promotional Price
    Each
       $869.75
    Supplier Number: F21585L