Order Entry
Canada
ContactUsLinkComponent
Methyl ethyl ketone ≥99%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Flammable
Irritant
Methyl ethyl ketone ≥99%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # CAMK620608
   Safer Human and Environmental Health
CAS Number:  
MOC:  Non-RegulatedCertificates
undefined
Flammable
Irritant
Methyl ethyl ketone ≥99%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog # CAMK620608
Supplier Number:  MACR6206-08
   Safer Human and Environmental Health
CAS Number:  
MOC:  Non-RegulatedCertificates

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Size:
    4 l
  • Environmentally Preferable:
    Safer Human and Environmental Health
  • Environmental certification:
    Chem21 Solvent selection guide recommended by default and after discussion
  • Cat. no.:
    CAMK620608
  • Supplier No.:
    6206-08

Specifications

Specification Test Results

  • ACS - Assay (CH₃COCH₂CH₃) (by GC):
    ≥99.0 %
  • ACS - Color:
    ≤15
  • ACS - Residue after Evaporation:
    ≤5 ppm
  • ACS - Water (H₂O)(by Karl Fischer titrn):
    ≤0.08 %
  • Optical Absorbance (1-cm path vs water) - 329 nm:
    ≤1.00
  • Optical Absorbance (1-cm path vs water) - 335 nm:
    ≤0.50
  • Optical Absorbance (1-cm path vs water) - 344 nm:
    ≤0.05
  • Optical Absorbance (1-cm path vs water) - 350 - 400 nm:
    ≤0.01
  • Titrable Acid (meq/g):
    ≤0.0005

Specification Test Results

About this item

 :
Merck Index:13,06097
Boiling Pt:79.6 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.8054 g/cm³ (20 °C)
Flash Pt:–4 °C
ADR:3,II
 :Management of Change (MOC) category = N
 :72.11 g/mol
Storage Temperature:Ambient
EINECS:201-159-0
UN:1193
REACH:01-2119457290-43
 :GHS02
 :GHS07
CAS Number:78-93-3
Formula:C₂H₅COCH₃
MDL Number:MFCD00011648