

Manganese(II) sulfate monohydrate 98.0-101.0%, powder, AR® ACS, Macron Fine Chemicals™
Catalog # CAMK619220
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number:
MOC: Non-RegulatedCertificates
Specifications
- Pack type:Case, Mall STAKMOR
- Size:12 kg
- Cat. no.:CAMK619220
Specifications
Specification Test Results
- ACS - Assay (MnSO₄·H₂O):98.0 - 101.0 %
- ACS - Calcium (Ca):≤0.005 %
- ACS - Chloride (Cl):≤0.005 %
- ACS - Heavy Metals (as Pb):≤0.002 %
- ACS - Insoluble Matter:≤0.01 %
- ACS - Iron (Fe):≤0.002 %
- ACS - Loss on Ignition:10.0 - 12.0 %
- ACS - Magnesium (Mg):≤0.005 %
- ACS - Nickel (Ni):≤0.02 %
- ACS - Potassium (K):≤0.01 %
- ACS - Sodium (Na):≤0.05 %
- Solubility:Passes Test
- ACS - Substances Reducing KMnO₄:Passes Test
- ACS - Zinc (Zn):≤0.005 %
Specification Test Results
About this item
:
Merck Index:13,05763
Melting Pt:700 °C
Density:2.95 g/cm³ (25 °C)
ADR:9,III
:Management of Change (MOC) category = N
:169.02 g/mol
EINECS:232-089-9
:GHS08
:GHS09
Storage Temperature:Refrigerator
CAS Number:10034-96-5
Formula:MnSO₄·1H₂O
MDL Number:MFCD00149159