Orders LinkContactUsLinkComponent
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Supplier:   Kemtech America
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Supplier:  Kemtech America
Certificates
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
48 Options Available Below
Product Details & Documents
Round-bottom flasks with one straight [ST] joint and one angled [ST] joint
  • Angled side neck
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Various sizes available
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Product Family Options
Product Information
  • Capacity
    Center neck [ST] joint
    Side neck ST joints
    Availability
    Price
  • 10 ml
    14/20
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $106.50
    $106.50
    New Customer Promotional Price
    Each
       $106.50
    Minimum order quantity is 1
    Supplier Number: F411110
  • 25 ml
    14/20
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $110.65
    $110.65
    New Customer Promotional Price
    Each
       $110.65
    Minimum order quantity is 1
    Supplier Number: F411125
  • 25 ml
    19/22
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $96.97
    $96.97
    New Customer Promotional Price
    Each
       $96.97
    Minimum order quantity is 1
    Supplier Number: F411925
  • 25 ml
    19/22
    19/22
    Log in for availability
    Log in for availability
    Each
    Each - $110.70
    $110.70
    New Customer Promotional Price
    Each
       $110.70
    Minimum order quantity is 1
    Supplier Number: F419925
  • 50 ml
    14/20
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $112.82
    $112.82
    New Customer Promotional Price
    Each
       $112.82
    Minimum order quantity is 1
    Supplier Number: F411150
  • 50 ml
    19/22
    19/22
    Log in for availability
    Log in for availability
    Each
    Each - $98.05
    $98.05
    New Customer Promotional Price
    Each
       $98.05
    Minimum order quantity is 1
    Supplier Number: F419950
  • 50 ml
    24/40
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $116.39
    $116.39
    New Customer Promotional Price
    Each
       $116.39
    Minimum order quantity is 1
    Supplier Number: F414150
  • 50 ml
    24/40
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $117.88
    $117.88
    New Customer Promotional Price
    Each
       $117.88
    Minimum order quantity is 1
    Supplier Number: F414450
  • 100 ml
    14/20
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $114.98
    $114.98
    New Customer Promotional Price
    Each
       $114.98
    Minimum order quantity is 1
    Supplier Number: F411100
  • 100 ml
    19/22
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $100.64
    $100.64
    New Customer Promotional Price
    Each
       $100.64
    Minimum order quantity is 1
    Supplier Number: F411900
  • 100 ml
    19/22
    19/22
    Log in for availability
    Log in for availability
    Each
    Each - $100.02
    $100.02
    New Customer Promotional Price
    Each
       $100.02
    Minimum order quantity is 1
    Supplier Number: F419900
  • 100 ml
    24/40
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $103.75
    $103.75
    New Customer Promotional Price
    Each
       $103.75
    Minimum order quantity is 1
    Supplier Number: F414100
  • 100 ml
    24/40
    19/22
    Log in for availability
    Log in for availability
    Each
    Each - $123.14
    $123.14
    New Customer Promotional Price
    Each
       $123.14
    Minimum order quantity is 1
    Supplier Number: F419100
  • 100 ml
    24/40
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $155.20
    $155.20
    New Customer Promotional Price
    Each
       $155.20
    Minimum order quantity is 1
    Supplier Number: F414400
  • 250 ml
    14/20
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $104.61
    $104.61
    New Customer Promotional Price
    Each
       $104.61
    Minimum order quantity is 1
    Supplier Number: F411250
  • 250 ml
    19/22
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $105.81
    $105.81
    New Customer Promotional Price
    Each
       $105.81
    Minimum order quantity is 1
    Supplier Number: F410250
  • 250 ml
    19/22
    19/22
    Log in for availability
    Log in for availability
    Each
    Each - $110.04
    $110.04
    New Customer Promotional Price
    Each
       $110.04
    Minimum order quantity is 1
    Supplier Number: F419250
  • 250 ml
    24/40
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $132.87
    $132.87
    New Customer Promotional Price
    Each
       $132.87
    Minimum order quantity is 1
    Supplier Number: F414251
  • 250 ml
    24/40
    19/22
    Log in for availability
    Log in for availability
    Each
    Each - $132.62
    $132.62
    New Customer Promotional Price
    Each
       $132.62
    Minimum order quantity is 1
    Supplier Number: F414259
  • 250 ml
    24/40
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $160.53
    $160.53
    New Customer Promotional Price
    Each
       $160.53
    Minimum order quantity is 1
    Supplier Number: F414250
  • 500 ml
    14/20
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $142.70
    $142.70
    New Customer Promotional Price
    Each
       $142.70
    Minimum order quantity is 1
    Supplier Number: F411501
  • 500 ml
    24/40
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $147.17
    $147.17
    New Customer Promotional Price
    Each
       $147.17
    Minimum order quantity is 1
    Supplier Number: F414501
  • 500 ml
    24/40
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $169.55
    $169.55
    New Customer Promotional Price
    Each
       $169.55
    Minimum order quantity is 1
    Supplier Number: F414504
  • 500 ml
    29/42
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $162.52
    $162.52
    New Customer Promotional Price
    Each
       $162.52
    Minimum order quantity is 1
    Supplier Number: F415501
  • 500 ml
    29/42
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $172.64
    $172.64
    New Customer Promotional Price
    Each
       $172.64
    Minimum order quantity is 1
    Supplier Number: F415502
  • 1000 ml
    24/40
    14/20
    Log in for availability
    Log in for availability
    Each
    Each - $174.48
    $174.48
    New Customer Promotional Price
    Each
       $174.48
    Minimum order quantity is 1
    Supplier Number: F41611L
  • 1000 ml
    24/40
    19/22
    Log in for availability
    Log in for availability
    Each
    Each - $173.66
    $173.66
    New Customer Promotional Price
    Each
       $173.66
    Minimum order quantity is 1
    Supplier Number: F41621L
  • 1000 ml
    24/40
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $207.40
    $207.40
    New Customer Promotional Price
    Each
       $207.40
    Minimum order quantity is 1
    Supplier Number: F41241L
  • 1000 ml
    29/42
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $203.57
    $203.57
    New Customer Promotional Price
    Each
       $203.57
    Minimum order quantity is 1
    Supplier Number: F41641L
  • 2000 ml
    24/40
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $289.43
    $289.43
    New Customer Promotional Price
    Each
       $289.43
    Supplier Number: F41242L
  • 2000 ml
    29/42
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $293.19
    $293.19
    New Customer Promotional Price
    Each
       $293.19
    Minimum order quantity is 1
    Supplier Number: F41712L
  • 2000 ml
    29/42
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $296.07
    $296.07
    New Customer Promotional Price
    Each
       $296.07
    Minimum order quantity is 1
    Supplier Number: F41722L
  • 2000 ml
    34/45
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $301.63
    $301.63
    New Customer Promotional Price
    Each
       $301.63
    Supplier Number: F41732L
  • 2000 ml
    34/45
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $301.63
    $301.63
    New Customer Promotional Price
    Each
       $301.63
    Minimum order quantity is 1
    Supplier Number: F41742L
  • 2000 ml
    45/50
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $308.12
    $308.12
    New Customer Promotional Price
    Each
       $308.12
    Supplier Number: F41752L
  • 2000 ml
    45/50
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $308.12
    $308.12
    New Customer Promotional Price
    Each
       $308.12
    Minimum order quantity is 1
    Supplier Number: F41762L
  • 3000 ml
    29/42
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $376.46
    $376.46
    New Customer Promotional Price
    Each
       $376.46
    Minimum order quantity is 1
    Supplier Number: F41813L
  • 3000 ml
    29/42
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $384.07
    $384.07
    New Customer Promotional Price
    Each
       $384.07
    Minimum order quantity is 1
    Supplier Number: F41823L
  • 3000 ml
    34/45
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $391.55
    $391.55
    New Customer Promotional Price
    Each
       $391.55
    Minimum order quantity is 1
    Supplier Number: F41833L
  • 3000 ml
    34/45
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $399.18
    $399.18
    New Customer Promotional Price
    Each
       $399.18
    Minimum order quantity is 1
    Supplier Number: F41843L
  • 3000 ml
    45/50
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $405.87
    $405.87
    New Customer Promotional Price
    Each
       $405.87
    Supplier Number: F41853L
  • 3000 ml
    45/50
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $411.74
    $411.74
    New Customer Promotional Price
    Each
       $411.74
    Supplier Number: F41863L
  • 5000 ml
    29/42
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $440.07
    $440.07
    New Customer Promotional Price
    Each
       $440.07
    Minimum order quantity is 1
    Supplier Number: F41915L
  • 5000 ml
    29/42
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $450.69
    $450.69
    New Customer Promotional Price
    Each
       $450.69
    Minimum order quantity is 1
    Supplier Number: F41925L
  • 5000 ml
    34/45
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $459.11
    $459.11
    New Customer Promotional Price
    Each
       $459.11
    Supplier Number: F41935L
  • 5000 ml
    34/45
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $468.22
    $468.22
    New Customer Promotional Price
    Each
       $468.22
    Minimum order quantity is 1
    Supplier Number: F41945L
  • 5000 ml
    45/50
    24/40
    Log in for availability
    Log in for availability
    Each
    Each - $475.20
    $475.20
    New Customer Promotional Price
    Each
       $475.20
    Minimum order quantity is 1
    Supplier Number: F41955L
  • 5000 ml
    45/50
    29/42
    Log in for availability
    Log in for availability
    Each
    Each - $484.33
    $484.33
    New Customer Promotional Price
    Each
       $484.33
    Minimum order quantity is 1
    Supplier Number: F41965L