


Phenol : chloroform : iso-amyl alcohol (25:24:1) ≥99.0%, OmniPur®, TE buffered saturated pH 6.7/8.0, Calbiochem®, Millipore®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:136112-00-0
:
MDL Number:MFCD00133763
Storage Temperature:Refrigerator
UN:2810
ADR:6.1,II
:GHS05
:GHS06
:GHS08
Specification Test Results
- Assay [phenol]:≥99.0%
- pH of Saturated phenol at 25°C:6.5-6.9
- pH of TRIS-EDTA buffer:7.8-8.2
- UV Absorbance at 330 nm:≤0.2AU
- UV Absorbance at 405 nm:≤0.1AU
- UV Absorbance at 510 nm:≤0.1AU
Specification Test Results
Related Information
Frequently Bought Together