



n-Pentane ≥99.9% (by GC), GC2™ for gas chromatography, for pesticide residue analysis, Burdick & Jackson™
Catalog # CABJGC312-4
Supplier: Burdick & Jackson
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:4 L
- Cat. no.:CABJGC312-4
- Supplier No.:GC312-4
Specifications
Specification Test Results
- Electron Capture GC (as lindane):2 ppt max.
- Flame Ionization GC (as nonane):1 ppb max.
- GC Analysis (n-Pentane):98 % min.
- GC Analysis (total C5 isomers):99.9 % min.
- Refractive Index (20ºC):1.3573 - 1.3579
- Residue:1 mg/L max.
- Water by Karl Fischer Titration:0.01 % max.
Specification Test Results
About this item
:
:72.15 g/mol
Merck Index:13,07193
Boiling Pt:7.8 %(V)
Melting Pt:–129.7 °C
Density:0.626 g/cm³ (20 °C)
Flash Pt:–49 °C
Storage Temperature:Ambient
EINECS:203-692-4
UN:1265
ADR:3,II
REACH:01-2119459286-30
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:109-66-0
Formula:H₃C(CH₂)₃CH₃
MDL Number:MFCD00009498