Urea ≥99% USP
About this item
GMP manufactured.
- GMP manufactured product
- CAS Number:57-13-6
- Formula:NH₂CONH₂
- MDL Number:MFCD00008022
- Molecular Weight:60.06 g/mol
- Merck Index:13,09935
- Boiling Pt:197 °C (1013 hPa)
- Melting Pt:132.7 °C
- Density:1.34 g/cm³ (20 °C)
- :Management of Change (MOC) category = R
- Storage Temperature:Ambient
- EINECS:200-315-5
- REACH:01-2119463277-33
4 Options Available Below
Product Details & Documents
GMP manufactured.
- GMP manufactured product
- CAS Number:57-13-6
- Formula:NH₂CONH₂
- MDL Number:MFCD00008022
- Molecular Weight:60.06 g/mol
- Merck Index:13,09935
- Boiling Pt:197 °C (1013 hPa)
- Melting Pt:132.7 °C
- Density:1.34 g/cm³ (20 °C)
- :Management of Change (MOC) category = R
- Storage Temperature:Ambient
- EINECS:200-315-5
- REACH:01-2119463277-33
Documents
Certificates
Search by Lot Number4206 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals Avantor Top Rated Guide
Product Literature
JTBaker_4206_Datasheet.pdf
Datasheet
Specifications for Product Family
Specification Test Results
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Assay (NH₂CONH₂):98.0 - 102.0 %
- USP - Residue on Ignition:≤0.1 %
- USP - Alcohol Insoluble Matter:≤0.04 %
- USP - Organic impurities: Urea related compound A:≤0.1 %
- USP - Organic impurities: Any individual unspecified impurity:≤0.1 %
- USP - Organic impurities: Total impurities:≤2.0 %
Specifications
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Discontinued-Supplier Number: -
Specifications
Cat. No.CAJT4206-60MOC: Regulated - Poly bottle, white, blue T/E cap500 gEach (500g)$640.67Case of 4 (500g)$1,729.31Supplier Number: BAKR4206-01
Specifications
Cat. No.CAJT4206-1MOC: Regulated - Poly bottle, white, blue T/E cap2.5 kgEach (3kg)$1,647.29Case of 4 (3kg)$4,373.95Supplier Number: BAKR4206-05
Specifications
Cat. No.CAJT4206-5MOC: Regulated - Supplier Number: BAKR4206-07
Specifications
Cat. No.CAJT4206-7MOC: Regulated - HDPE drum, Flowmor™ package50 kgDiscontinued-Supplier Number: -
Specifications
Cat. No.CAJT4206-60MOC: Regulated - Supplier Number: BAKR4206-F8
Specifications
Cat. No.CAJT4206-F8MOC: Regulated - Fiber drum, Leverpack®110 lbs.Discontinued-Supplier Number: -
Specifications
Cat. No.CAJT4206-9MOC: Regulated
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



