Order Entry
Canada
ContactUsLinkComponent
Sodium Cacodylate, 0.1 M Buffer Solution, pH 6.8
Sodium Cacodylate, 0.1 M Buffer Solution, pH 6.8
Catalog # CAAAJ62202-AE
CAS Number:  
Sodium Cacodylate, 0.1 M Buffer Solution, pH 6.8
Catalog # CAAAJ62202-AE
Supplier Number:  J62202-AE
CAS Number:  

Specifications

  • CAS No.:
    6131-99-3
  • Size:
    100 ml
  • MDL No.:
    MFCD00149079
  • Storage temperature:
    2...8 °C
  • Physical form:
    Liquid
  • Cat. no.:
    CAAAJ62202-AE
  • Supplier No.:
    J62202-AE

Specifications

About this item

Liquid

CAS No.: 6131-99-3
Storage Temperature: 2 to 8 ℃
Physical Form: Liquid
MDL No.: MFCD00149079