ContactUsLinkComponent
Symmetrical Round Jaws Double Buret Clamps
Catalog # 470005-374
Supplier:  GSC International
undefined
Symmetrical Round Jaws Double Buret Clamps
Catalog # 470005-374
Supplier:  GSC International
Supplier Number:  4-BCDR86
Specifications
  • Description:
    Clamp, buret, double, with symmetrical round rubber coated jaws, Humboldt
  • Environmentally Preferable:
  • Cat. no.:
    470005-374
  • Supplier No.:
    7-G86
Specifications
About this item
Use the two sets of symmetrical round jaws for simultaneously holding two round objects, each up to 35 mm in diameter. Each of the rubber-coated clamps rotates 360° and may be locked at any angle. The unit attaches to any standard support rod up to 13 mm in diameter.