Orders LinkContactUsLinkComponent
Specific Gravity Hydrometers with Thermometer
Specific Gravity Hydrometers with Thermometer
Supplier:   Heathrow Scientific
Specific Gravity Hydrometers with Thermometer
Certificates
Specific Gravity Hydrometers with Thermometer
Specific Gravity Hydrometers with Thermometer
3 Options Available Below
Product Details & Documents
These specific gravity hydrometers feature an internal thermometer for calculating temperature corrections and are calibrated for use at 60 ℉.
  • The liquid fill of these internal thermometers is safety blue
The precision specific gravity hydrometer feature and internal thermometer for calculating temperature corrections.
Product Family Options
Product Information
  • Temperature range
    Range
    Length
    Subdivisions
    Availability
    Price
  • 20...150 °F
    1.000...1.070
    385 mm (15")
    Hydrometer: 0.001
    Thermometer: 2 °F
    Log in for availability
    Log in for availability
    Each
    Each - $88.02
    $88.02
    New Customer Promotional Price
    Each
       $88.02
    Supplier Number: 6602TS-7
  • 20...150 °F
    1.060...1.130
    385 mm (15")
    Hydrometer: 0.001
    Thermometer: 2 °F
    Log in for availability
    Log in for availability
    Each
    Each - $88.02
    $88.02
    New Customer Promotional Price
    Each
       $88.02
    Supplier Number: 6602TS-8
  • 20...150 °F
    1.000...1.220
    305 mm (12")
    Hydrometer: 0.002
    Thermometer: 2 °F
    Log in for availability
    Log in for availability
    Each
    Each - $91.66
    $91.66
    New Customer Promotional Price
    Each
       $91.66
    Supplier Number: 6603TS-1