Orders LinkContactUsLinkComponent
Synthware Flask, Filtering, Microscale, Kemtech America
Synthware Flask, Filtering, Microscale, Kemtech America
Synthware Flask, Filtering, Microscale, Kemtech America
Synthware Flask, Filtering, Microscale, Kemtech America
Filtering flask for microscale applications.
  • Microscale
  • Heavy wall Borosilicate construction
  • Beaded top rim
  • Barbed side arm
Top is tooled for a #2 neoprene filter adapter or #3 rubber stopper.
Product Family Options
  • Capacity
  • 25 ml
    Each
    Each - $47.05
    $47.05
    Each
       $47.05
     : F150025
  • 50 ml
    Each
    Each - $47.05
    $47.05
    Each
       $47.05
     : F150050