ContactUsLinkComponent
TAE Buffer, Concentrate, OmniPur*. 25X.
TAE Buffer, Concentrate, OmniPur*. 25X.
Catalog # CAEM8750
Supplier:  MilliporeSigma
CAS Number:  
TAE Buffer, Concentrate, OmniPur*. 25X.
Catalog # CAEM8750
Supplier:  MilliporeSigma
Supplier Number:  8750-1PC
CAS Number:  
Specifications
  • Pack type:
    Poly bag
  • Size:
    1 L
  • Cat. no.:
    CAEM8750
  • Supplier No.:
    8750
Specifications
About this item
Premixed powder
Each size makes the specified volume of 25X TAE (1X=0.04M Tris, 0.04M Acetate, 0.001M EDTA). DNase-, RNase-, and protease-free.