ContactUsLinkComponent
BRAND® PCR Plates, 96-Well, Rigid Frame, BIO-CERT® PCR QUALITY
BRAND® PCR Plates, 96-Well, Rigid Frame, BIO-CERT® PCR QUALITY
  77447-860
 :  Brandtech
undefined
BRAND® PCR Plates, 96-Well, Rigid Frame, BIO-CERT® PCR QUALITY
  77447-860
 :  Brandtech
 :  781541
  • Frame Color:
    Frosted
  • No. of wells:
    96
  • Profile:
    Low
  • Skirt:
    Fully skirted
  • Well Color:
    Transparent
  • Well Volume:
    0.15 ml
  • Description:
    PCR rigid frame plate, 96-well, H1 cut corner
  • Cat. no.:
    77447-860
BRAND® 96-well plates provide efficient PCR and qPCR solutions for medium to high sample throughput, featuring a strong polycarbonate frame and thin-walled wells for rapid temperature transfer.
  • Compatible with all standard thermal cyclers and fully automation ready
  • Select from semi-skirted or fully skirted plate options
  • Distinguish workflow steps easily with a range of color choices
  • White plates for qPCR, optimized to reflect fluorescence signals
BRAND® rigid frame PCR plates feature a robust polycarbonate frame, ensuring safe handling in both semi-automated and automated pipetting systems while providing optimal evaporation protection.
The rigid frame design also facilitates the use of ultra-thin-walled wells made from 100% virgin medical-grade polypropylene, ensuring rapid temperature transfer from the cycler to the sample, consequently reducing cycle times. The 96-well rigid frame plates are manufactured in controlled cleanroom environments.
 : BIO-CERT® PCR quality level, made from virgin, raw polypropylene, free of human DNA, RNase, DNase, pyrogens, and PCR-inhibitors.