Sodium Chloride Hydrometers
Supplier: Heathrow Scientific
Certificates
About this item
These hydrometers measure the percentage of sodium chloride. Specifically, model 6611-1 measures the saturation percentage of sodium chloride in solutions and model 6611-2 measures the percentage by weight of sodium chloride in solutions.
- 30.5 cm (12") length
Both models are calibrated for use at 15 °C (60 °F).
Customers Who Bought This Also Bought