ContactUsLinkComponent
KIMAX® Low Form Starter Packs
KIMAX® Low Form Starter Packs
Catalog # 89209-302
Supplier #: 14085-01
undefined
KIMAX® Low Form Starter Packs
Catalog # 89209-302
Supplier Number:  14080-01
Supplier #: 14085-01
Log in for availability
Select an Option
Includes 1 each 250 ml, 400 ml, 600 ml and 1 L
Select an Option
Low form, heavy duty beaker starter pack
Select an Option
89209-302
Select an Option
14085-01
About this item
An assortment of popularly sized “variety pack” glass beakers from our 14000 Low Form Griffin series that is ideal for start-up labs and customers who need a variety of beaker sizes but have limited lab space or glassware needs.
  • Double capacity scale
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Frequently Bought Together