- Description:Rapid Response® Strep A Rapid Test Strip
- Size:25 Tests/Kit
- Cat. no.:CA77448-060
The Rapid Response® Strep A Rapid Test Device is a rapid test for the presumptive detection of Group A Streptococcus antigens in human throat swab specimens. This kit is intended for use as an aid in the diagnosis of Strep A infection directly from throat swabs to help diagnose and administer therapy immediately.
- Sample: Throat swab
- Format: Strip
- Rapid test for detecting Strep A infection
- Controls included in kit
- Includes all materials needed to perform the test
- Comes with a tube stand for mess-free workspace
- Read results in 5 minutes
The Rapid Response® Strep A Rapid Test Device is a rapid visual immunoassay that detects Strep A in throat swabs to help diagnose and administer therapy treatment immediately.
Beta-hemolytic Group A Streptococcus is a major cause of upper respiratory infections such as tonsillitis, pharyngitis, and scarlet fever. Early diagnosis and treatment has been shown to reduce the severity of symptoms and further complications, such as rheumatic fever and glomerulonephritis. Conventional methods for detecting Strep A infection are dependent on isolation and subsequent identification of the organism, and often require 24-48 hours. With Rapid Response® Strep A Rapid Test Device, get results in just 5 minutes from a quick throat swab.
Each kit contains individually packaged test strips. Reagent bottles, sterile swabs, extraction tubes, nozzles with filter, controls, a tube stand, and a detailed instructions are included in every kit.
To perform the test, place an extraction tube in the tube stand and add 4 drops of reagent 1 and reagent 2 to the tube. Mix the solution and immediately immerse the swab with the collected throat sample into the tube. Let stand for 1-2 minutes and then remove the swab. Attach the nozzle onto the tube. Add 3 drops of extracted solution onto the sample region on the test strip and then read the results at 5 minutes. Interpretation of results are: 2 colored lines one in control region (C) and another in test region (T) is positive, 1 line in the control region (C) and no line in the test region (T) is negative, no lines or no line in the Control Region (C) is invalid.