Order Entry
Canada
ContactUsLinkComponent
BD® Thioglycollate Medium, Double Wrapped, BD Diagnostics
BD® Thioglycollate Medium, Double Wrapped, BD Diagnostics
Catalog # CA90003-530
Supplier:  BD
CAS Number:  
undefined
BD® Thioglycollate Medium, Double Wrapped, BD Diagnostics
Catalog # CA90003-530
Supplier:  BD
Supplier Number:  257217
CAS Number:  

Specifications

  • Pack type:
    Bottle, infusion type
  • Description:
    BD Fluid thioglycollate medium
  • Cat. no.:
    CA90003-530
  • Cap Type:
    Septum
  • Fill Volume:
    100 ml
  • Volume:
    125 ml
  • Supplier No.:
    257217

Specifications

About this item