Sterile PETG Square Graduated Media Bottles
Supplier: Qorpak
New Product
Certificates
About this item
Ergonomic Media bottles are suited for biopharmaceutical processing, storage of critical reagents, media, and various buffers used in laboratories. Packaging is designed for cleanroom applications.
- Made of USP Class VI raw material, with low leachables and extractables
- Manufactured in a Class 8 cleanroom
- Non-pyrogenic, non-cytotoxic, endotoxin-free, and nuclease-free
- Gamma sterile product
- Graduated for quick user visibility
The bottle and closure are secured with a tamper-resistant, heat-shrink band, and the bottle body is square-shaped, making it convenient for storage purposes. The bottle surface is smooth and shiny, easy to clean, and does not easily leave a residue. The resin materials are chosen to minimize additives and potential leachables. No fillers or plasticizers are used.
Customers Who Bought This Also Bought