Formaldehyde solution 37% stabilized, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
MOC: Non-Regulated
About this item
CAS Number:50-00-0
:
Formula:CH₂O
MDL Number:MFCD00003274
:30.03 g/mol
Density:1.08 g/cm³
Flash Pt:62
Storage Temperature:Ambient
EINECS:200-001-8
UN:1198
ADR:3(8),III
:Stabilised with methanol 10-15 %
:GHS02
:GHS05
:GHS06
:GHS08
Specification Test Results
- ACS - Assay (HCHO) (by acidimetry):36.5 - 38.0 %
- ACS - Color (APHA):≤10
- ACS - Residue after Ignition:≤0.005 %
- ACS - Titrable Acid (meq/g):≤0.006
- ACS - Sulfate (SO₄):≤0.002 %
- ACS - Preservative (methanol):10 - 12 %
- Chloride (Cl):≤5 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤2.0 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number2106 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals Avantor Top Rated Guide
Product Literature
Brand Definition Baker Analyzed
Datasheet
sds_BAKR2106
SDS
Frequently Bought Together