


Diethyl ether ≥98% (by GC) stabilized, AR® ACS, Macron Fine Chemicals™
Supplier: Avantor
MOC: Non-Regulated
About this item
CAS Number:60-29-7
:
Formula:(CH₃CH₂)₂O
MDL Number:MFCD00011646
:74.12 g/mol
Merck Index:12,03852
Boiling Pt:35 °C (1013 hPa)
Melting Pt:–123 °C
Density:0.7135 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:3,I
:Management of Change (MOC) category = N
Flash Pt:–40 °C
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
EINECS:200-467-2
UN:1155
REACH:01-2119535785-29
:GHS02
:GHS07
:GHS08
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- ACS - Assay ((C₂H₅)₂O) (by GC, exclusive of preservative):≥98.0 %
- Appearance (clear, colorless liquid):Passes Test
- ACS - Carbonyl Compounds (as HCHO) (by polarography):≤0.001 %
- ACS - Color (APHA):≤10
- ACS - Peroxide (as H₂O₂):≤1 ppm
- ACS - Preservative (BHT):≥0.00005 %
- ACS - Residue after Evaporation:≤0.001 %
- ACS - Titrable Acid (µeq/g):≤0.2
Specification Test Results
Related Information
Certificates
Search by Lot Number0850 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_MACR0850
SDS
Frequently Bought Together