Order Entry
Canada
ContactUsLinkComponent
Ethidium bromide 10 mg/mL in aqueous solution, OmniPur®, Millipore®
Toxic
Health
Ethidium bromide 10 mg/mL in aqueous solution, OmniPur®, Millipore®
Catalog # CA-EM4410
Supplier:  MilliporeSigma
CAS Number:  
Toxic
Health
Ethidium bromide 10 mg/mL in aqueous solution, OmniPur®, Millipore®
Catalog # CA-EM4410
Supplier:  MilliporeSigma
Supplier Number:  4410-10ML
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    10 mL
  • Cat. no.:
    CA-EM4410
  • Supplier No.:
    4410

Specifications

About this item

 :
 :394.31 g/mol
EINECS:214-984-6
 :GHS06
 :GHS08
CAS Number:1239-45-8
Formula:C₂₁H₂₀BrN₃
MDL Number:MFCD00011724