Order Entry
Canada
ContactUsLinkComponent
QuantiFluo™ Adenosine Deaminase Assay Kit
QuantiFluo™ Adenosine Deaminase Assay Kit
Catalog # 77441-646
Supplier:  BIOASSAY SYSTEMS
CAS Number:  
undefined
QuantiFluo™ Adenosine Deaminase Assay Kit
Catalog # 77441-646
Supplier:  BIOASSAY SYSTEMS
Supplier Number:  DADA-100
CAS Number:  

Specifications

  • Description:
    QuantiFluo™ adenosine deaminase assay kit
  • Size:
    100 tests
  • Cat. no.:
    77441-646

Specifications

About this item

For quantitative determination of adenosine deaminase enzyme activity in biological samples.

  • Fast and safe
  • Assay can be completed within 50 minutes
  • Nonradioactive assay
  • Sensitive and accurate
  • Linear detection range is 0.3 to 30 U/L
  • adenosine deaminase in a 96-well plate assay
  • Convenient and high-throughput
  • Homogeneous 'mix-incubatemeasure' type assay
  • Can be readily automated to assay thousands of samples per day

Adenosine deaminase (EC 3.5.4.4) is a key enzyme in purine metabolism and plays an important role in the normal immune function of humans. ADA deficiency is one cause of Severe Combined Immunodeficiency (SCID), and elevated levels of ADA have been observed in patients with tuberculosis and immune-related diseases.

BioAssay systems DADA-100 Kit provides a convenient fluorimetric method to measure adenosine deaminase activity in biological samples. In this assay, liberated ammonia reacts with the reagents where the increase in fluorescence at λex/em = 415/475 nm is directly proportional to enzyme activity.