1,6-Dibromohexane ≥97.0%
Product Details & Documents
CAS Number: 629-03-8
MDL Number: MFCD00000272
Molecular Formula: C6H12Br2
Molecular Weight: 243.97
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 246
Melting point (°C): -2
Flash Point (°C): 152
Specific Gravity (20/20): 1.61
Other Product Names / Synonyms: Hexamethylene Bromide, Hexamethylene Dibromide
- CAS Number:629-03-8
- Formula:BrH₂C(CH₂)₄CH₂Br
- MDL Number:MFCD00000272
- Molecular Weight:243.97 g/mol
- Boiling Pt:246 °C
- Melting Pt:–2 °C
- Density:1.61 (20 °C)
- Flash Pt:152 °C
- Storage Temperature:Ambient
- EINECS:211-067-2
- UN:3082
- ADR:9,III
Documents
Specifications
Specification Test Results
- Purity/Analysis Method:97.0% (GC)
Specifications
- Size:100 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



