


N,N-Dimethylformamide ≥99.8% (by GC, corrected for water content), BakerDRY™ for organic synthesis, J.T.Baker®
Supplier: VWR International
MOC: Non-Regulated, With Information
About this item
CAS Number:68-12-2
:
Formula:HCON(CH₃)₂
MDL Number:MFCD00003284
:73.09 g/mol
Merck Index:13,03269
Boiling Pt:153 °C (1013 hPa)
Melting Pt:–61 °C
Density:0.949 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-679-5
UN:2265
ADR:3,III
:Management of Change (MOC) category = NI
Flash Pt:57.5 °C (closed cup)
REACH:01-2119475605-32
:GHS02
:GHS08
:GHS07
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- ACS - Assay (HCON(CH₃)₂) (by GC, corrected for water):≥99.8 %
- Appearance:Passes Test
- ACS - Color (APHA):≤15
- ACS - Residue after Evaporation:≤0.005 %
- ACS - Titrable Acid (meq/g):≤0.0005
- ACS - Titrable Base (meq/g):≤0.003
- ACS - Water (H₂O)(by Karl Fischer titrn):≤20 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number9213 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
BakerDry Grade Definition
Datasheet
Frequently Bought Together