Dimethyl sulfoxide ≥99.9% (by GC, corrected for water content), PHOTREX® for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
CAS Number:67-68-5
:
Formula:(CH₃)₂SO
MDL Number:MFCD00002089
:78.14 g/mol
Merck Index:13,03285
Boiling Pt:189 °C (1013 hPa)
Melting Pt:18.5 °C
Density:1.101 g/cm³ (20 °C)
Flash Pt:87 °C (closed cup)
ADR:4,
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Environmentally Preferable:Safer Human and Environmental Health
- Environmental certification:Chem21 solvent selection guide recommended by default
Specifications
Specification Test Results
- Appearance:Passes Test
- ACS - Assay ((CH₃)₂SO) (by GC, corrected for water):≥99.9 %
- ACS - Residue after Evaporation:≤0.002 %
- ACS - Titrable Acid (meq/g):≤0.001
- ACS - Water (by KF, coulometric):≤0.1 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 400-350:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 330 nm:≤0.02
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 310 nm:≤0.06
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 290 nm:≤0.18
- Ultraviolet Absorbance (1.00-cm cell vs. water) (curve smooth throughout stated range with no extraneous impurity peaks) - 270 nm:≤0.40
Specification Test Results
Related Information
Certificates
Search by Lot Number9194 Product Specifications
Additional Product Details
VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
Frequently Bought Together