Bovine Desoxyribonuclease I (DNase I) (from Pancreas)
CA3750
:
- Pack type:Glass vial
- CAS No.:9003-98-9
- Source:Pancreas
- Species:Bovine
- Size:1 vial
- Storage conditions:Freezer
- Enzyme name:Desoxyribonuclease I (DNase I)
- Cat. no.:CA3750
- Supplier No.:3750
White to off-white crystalline powder
Enzyme. For molecular biology. RNase-free. Store frozen at –15 to –22°C. 50,000 units per vial.