

Dichloromethane ≥99.9% stabilised, B&J Brand™ ACS for HPLC, for spectrophotometry, Burdick & Jackson™
Supplier: Burdick & Jackson
Certificates
About this item
CAS Number:75-09-2
:
Formula:CH₂Cl₂
MDL Number:MFCD00000881
:84.93 g/mol
Merck Index:12,06140
Boiling Pt:40 °C
Melting Pt:–95 °C
Density:1.33 g/cm³
Flash Pt: Does not flash
UN:1593
ADR:6.1,III
:Stabilised with amylene
EINECS:200-838-9
:GHS07
:GHS08
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Acidity (as HCl):1 mg/L max.
- Chloride:10 mg/L max.
- Electron Capture GC:10 ng/L max.
- Flame Ionization GC (as nonane):5 ppb max.
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.4236 - 1.4246
- Residue:1 mg/L max.
- UV Cutoff:233 nm max.
- Water by Karl Fischer Titration:0.01 % max.
Specification Test Results
Related Information
Frequently Bought Together