TouchNTuff® 92-600, Nitrile Gloves, Powder-Free, Ansell
Supplier: Ansell Healthcare
CE Compliant
Certificates
About this item
The TouchNTuff 92-600 gloves are thin (4 mil), 100% nitrile disposable gloves which are applicable for chemical splash protection.
- Superior puncture resistance
- Exceptional durability and fingertip sensitivity
- Silicone-free
They offer three times the puncture resistance of standard latex or vinyl gloves. These gloves combine excellent fingertip sensitivity with exceptional durability to give a high level of protection against grease and oil. They are lightweight and comfortable to wear.
Certifications: Touch N Tuff® gloves comply with FDA food handling requirements (21 CFR 177.2600).
Specifications
- Catalog No:
- CA76006-392
- CA76006-394
- CA76006-390
- CA34800-502
- CA34800-500
- CA34800-506
- CA34800-504
- CA76006-396
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
Specifications
Related Information
Certificates
Search by Lot Number34800-500_19-0928
Conformity declaration
Focus Safety Volume 1 Issue 1 Safety Essentials Volume 2
Product Literature
Customers Who Bought This Also Bought