Nitrilite® 93-401, Silky Ultra-Clean, Gloves, Ansell
Supplier: Ansell Healthcare
Certificates
About this item
These ambidextrous gloves have the superior strength of nitrile yet are thinner than comparable vinyl or latex gloves for a lighter, more comfortable glove with improved dexterity and tactility.
- Beaded cuffs
- Powder-free
- Strong, yet light and flexible
These gloves significantly reduce particlate and extractable levels.
Ideal applications include electronics assembly, handling sensitive parts and materials, pharmaceutical manufacturing and product protection in handling cosmetics, packaging, optical goods, microelectronic and micro-mechanic components, as well as protection from blood and other bodily fluids.
They are powder-free with no plasicizers, silicone or pigments.
Ordering information: Gloves are packaged in an ISO Class 5 (FED-STD-209E Class 100/M3.5) environment. Cleanroom-compatible packaging contains 50 gloves per inner poly bag and 2 inner bags per outer poly bag. Lot number is printed on outer poly bag in IPA-resistant ink.
Specifications
- Catalog No:
- CA47748-238
- CA47748-232
- CA47748-234
- CA47748-236
- CA80093-140
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Textured Fingertips
- Textured Fingertips
- Textured Fingertips
- Textured Fingertips
- Textured Fingertips
- Finger thickness:
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- 6 mil
- Palm thickness:
- 5 mil
- 5 mil
- 5 mil
- 5 mil
- 5 mil