Calcium Chloride Standard, 1 ml = 1 mg CaCO₃ (0.4 mg Ca), 1000 ppm CaCO₃v (400 ppm Ca), 0.0100 M Ca2+, 0.0200 N Ca2+, Ricca Chemical Company
Catalog # RC178016
Supplier: Ricca Chemical
Small Business Enterprise
CAS Number:
Specifications
- Concentration:400 µg/ml Ca
- Matrix:H₂O/tr. HCl
- Pack type:Poly Bottle
- Size:16 oz.
- Cat. no.:RC178016
- Element:Calcium
- Supplier No.:R1780000-500A
Specifications
About this item
APHA and ASTM
1mL = 1mg CaCO3.