ComfortGrip® Latex Gloves, Microflex®, Ansell
Supplier: Microflex
Certificates
About this item
Disposable, medical-grade examination gloves are made of natural rubber latex, which offers excellent barrier integrity and elasticity for comfortable hand protection.
- Powder-free natural rubber latex
- Ambidextrous
- Fully textured, beaded cuff
These low-protein gloves contain ≤50 µg of total water-extractable protein per gram.
Certifications: Gloves meet or exceed ASTM D3578 standards.
Caution: This product contains natural rubber latex, which may cause allergic reactions. Safe use of this glove by or on latex-sensitive individuals has not been established.
Specifications
- Catalog No:
- CA14900-534
- CA14900-532
- CA14900-540
- CA14900-538
- CA14900-536
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff thickness:
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- Finger thickness:
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- Palm thickness:
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil
- 5.1 mil