Order Entry
Canada
ContactUsLinkComponent
Anti-SARS-CoV-2 (2019-nCoV) Spike protein S2 Rabbit Polyclonal Antibody
Anti-SARS-CoV-2 (2019-nCoV) Spike protein S2 Rabbit Polyclonal Antibody
Catalog # 77440-178
Supplier:  Bioss
Anti-SARS-CoV-2 (2019-nCoV) Spike protein S2 Rabbit Polyclonal Antibody
Catalog # 77440-178
Supplier:  Bioss
Supplier Number:  BS-24425R

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    SARS-CoV-2 (2019-nCoV) Spike protein S2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG (H, L)
  • Reactivity:
    SARS-CoV-2
  • Western blot:
    Yes
  • Size:
    100 µl
  • Gene ID:
    43740568
  • Storage buffer:
    0.01 M TBS (pH 7.4) with 1% BSA, 0.02% Proclin300 and 50% glycerol
  • Molecular weight:
    141/65
  • Storage temperature:
    Shipped at 4 ℃. Store at −20 °C for one year. Avoid repeated freeze/thaw cycles.
  • Concentration:
    1 µg/µl
  • Purification:
    Affinity purified by protein A
  • Cat. no.:
    77440-178
  • Packaging:
    Vial

Specifications

About this item

The spike (S) glycoprotein of coronaviruses contains protrusions that will only bind to certain receptors on the host cell. Known receptors bind S1 are ACE2, angiotensin-converting enzyme 2; DPP4, dipeptidyl peptidase-4; APN, aminopeptidase N; CEACAM, carcinoembryonic antigen-related cell adhesion molecule 1; Sia, sialic acid; O-ac Sia, O-acetylated sialic acid. The spike is essential for both host specificity and viral infectivity. The term 'peplomer' is typically used to refer to a grouping of heterologous proteins on the virus surface that function together. The spike (S) glycoprotein of coronaviruses is known to be essential in the binding of the virus to the host cell at the advent of the infection process. It's been reported that 2019-nCoV can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity. The main functions for the Spike protein are summarized as: Mediate receptor binding and membrane fusion; Defines the range of the hosts and specificity of the virus; Main component to bind with the neutralizing antibody; Key target for vaccine design; Can be transmitted between different hosts through gene recombination or mutation of the receptor binding domain (RBD), leading to a higher mortality rate.

Type: Primary
Antigen: SARS-CoV-2 (2019-nCoV) Spike protein S2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (H, L)
Reactivity: SARS-CoV-2